This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03075314_m1
See other RGS14A GE Assays ›
Gene Symbol:
rgs14a
Gene Name:
regulator of G protein signaling 14a
Gene Aliases:
rgs14, wu:fi04g12
Chromosome Location:
Chr.14: 872012 - 899155 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03075314_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rgs14a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_201304.1 NP_958461.1 12 - 13 1498 68
XM_009307644.3 XP_009305919.2 12 - 13 1464 68
GenBank mRNA BC055659.1 - 12 - 13 1498 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

spindle organization
signal transduction
germ cell migration
intracellular signal transduction
cell division
G-protein alpha-subunit binding
GDP-dissociation inhibitor activity
GTPase activator activity
microtubule binding
GTPase regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0