This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03074231_m1
See other PTK2AB GE Assays ›
Gene Symbol:
ptk2ab
Gene Name:
protein tyrosine kinase 2ab
Gene Aliases:
fak, fak1a, ptk2, ptk2.1, ptk2a, wu:fa18c04, zgc:66114
Chromosome Location:
Chr.16: 30737278 - 30869776 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03074231_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ptk2ab
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131796.1 NP_571871.1 3 - 4 407 131
XM_005158162.4 XP_005158219.1 2 - 3 783 131
XM_005158163.4 XP_005158220.1 2 - 3 783 131
XM_005158164.4 XP_005158221.1 2 - 3 783 131
XM_005158165.4 XP_005158222.1 2 - 3 783 131
XM_009292411.3 XP_009290686.1 2 - 3 783 131
XM_021466598.1 XP_021322273.1 2 - 3 785 131
XM_021466599.1 XP_021322274.1 2 - 3 327 131
XM_021466600.1 XP_021322275.1 2 - 3 785 131
XM_021466601.1 XP_021322276.1 2 - 3 784 131
XM_021466602.1 XP_021322277.1 3 - 4 349 131
XM_021466603.1 XP_021322278.1 2 - 3 785 131
XM_021466604.1 XP_021322279.1 2 - 3 784 131
XM_021466605.1 XP_021322280.1 2 - 3 784 131
XM_021466606.1 XP_021322281.1 2 - 3 784 131
XM_021466607.1 XP_021322282.1 2 - 3 784 131
XM_021466608.1 XP_021322283.1 2 - 3 783 131
XM_021466609.1 XP_021322284.1 3 - 4 444 131
GenBank mRNA AF348085.1 - 3 - 4 407 131

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
signal complex assembly
epidermal growth factor receptor signaling pathway
phosphorylation
midbrain-hindbrain boundary morphogenesis
cell differentiation
regulation of cell adhesion
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
innate immune response
lens fiber cell morphogenesis
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor binding
ATP binding
kinase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0