This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03074004_m1
See other APOA1A GE Assays ›
Gene Symbol:
apoa1a
Gene Name:
apolipoprotein A-Ia
Gene Aliases:
apoa, apoa1, cb49, wu:fb33f01, zgc:103718
Chromosome Location:
Chr.5: 142014 - 145025 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03074004_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: apoa1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131128.1 NP_571203.1 3 - 4 210 92
GenBank mRNA Y13653.1 - 210 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

apolipoprotein

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
cholesterol biosynthetic process
lipid transport
steroid metabolic process
cholesterol metabolic process
positive regulation of cholesterol esterification
cholesterol transport
cholesterol efflux
phospholipid efflux
very-low-density lipoprotein particle remodeling
high-density lipoprotein particle assembly
lipoprotein metabolic process
cholesterol homeostasis
reverse cholesterol transport
positive regulation of lipid biosynthetic process
triglyceride homeostasis
lipid transporter activity
phospholipid binding
high-density lipoprotein particle binding
lipid binding
cholesterol binding
phosphatidylcholine-sterol O-acyltransferase activator activity
intermembrane cholesterol transfer activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-867854cbc7-vwcfx:80/100.66.130.94:80.
git-commit: 25ccc7b1e9fd8ebf0690872e1fef11e169411d8b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0