This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03073758_g1
See other RYR1B GE Assays ›
Gene Symbol:
ryr1b
Gene Name:
ryanodine receptor 1b (skeletal)
Gene Aliases:
im:7137738, ryrab, si:dkey-250i19.1
Chromosome Location:
Chr.18: 35489483 - 35736799 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03073758_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ryr1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001102571.1 NP_001096041.1 67 - 68 9756 62
XM_017351957.2 XP_017207446.1 68 - 69 9985 62
XM_017351958.2 XP_017207447.1 68 - 69 9985 62
XM_017351959.2 XP_017207448.1 68 - 69 9985 62
XM_017351960.2 XP_017207449.1 68 - 69 9985 62
XM_017351961.2 XP_017207450.1 68 - 69 9985 62
XM_017351962.2 XP_017207451.1 67 - 68 9964 62
XM_017351963.2 XP_017207452.1 67 - 68 9964 62
XM_021467693.1 XP_021323368.1 68 - 69 9985 62
XM_021467694.1 XP_021323369.1 68 - 69 9985 62
XM_021467695.1 XP_021323370.1 67 - 68 9964 62
XM_021467696.1 XP_021323371.1 68 - 69 9985 62
GenBank mRNA AB247454.1 - 67 - 68 9756 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

skeletal muscle contraction
ion transport
calcium ion transport
cellular calcium ion homeostasis
larval locomotory behavior
fast-twitch skeletal muscle fiber contraction
slow-twitch skeletal muscle fiber contraction
cellular response to oxidative stress
release of sequestered calcium ion into cytosol
transmembrane transport
calcium ion transmembrane transport
ion channel activity
ryanodine-sensitive calcium-release channel activity
calcium channel activity
calcium ion binding
protein binding
calcium-induced calcium release activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0