This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03073385_m1
See other PRKACAA GE Assays ›
Gene Symbol:
prkacaa
Gene Name:
protein kinase, cAMP-dependent, catalytic, alpha, genome duplicate a
Gene Aliases:
zgc:158799
Chromosome Location:
Chr.3: 49455959 - 49504023 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03073385_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: prkacaa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001082840.1 NP_001076309.1 2 - 3 680 80
XM_021470955.1 XP_021326630.1 2 - 3 439 80
GenBank mRNA BC133159.1 - 2 - 3 680 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
protein kinase A signaling
phosphorylation
cellular response to glucose stimulus
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
cAMP-dependent protein kinase activity
ATP binding
kinase activity
transferase activity
protein kinase A regulatory subunit binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0