This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03072313_m1
See other SLC9A7 GE Assays ›
Gene Symbol:
slc9a7
Gene Name:
solute carrier family 9 member 7
Gene Aliases:
NHE7, wu:fk03b09, zgc:101088, zgc:113878
Chromosome Location:
Chr.6: 37529381 - 37600648 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03072313_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slc9a7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030077.2 NP_001025248.2 6 - 7 939 60
XM_005165986.4 XP_005166043.1 6 - 7 940 60
XM_009302520.3 XP_009300795.1 6 - 7 941 60
GenBank mRNA BC096982.1 - 6 - 7 939 60
BC165247.1 - 6 - 7 895 60
EF591987.1 - 6 - 7 936 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

ion transport
cation transport
sodium ion transport
regulation of pH
regulation of intracellular pH
transmembrane transport
potassium ion transmembrane transport
sodium ion import across plasma membrane
antiporter activity
solute:proton antiporter activity
sodium:proton antiporter activity
potassium:proton antiporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0