This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03070972_m1
See other CEP43 GE Assays ›
Gene Symbol:
cep43
Gene Name:
centrosomal protein 43
Gene Aliases:
ccr6a, fgfr1op, zgc:86782
Chromosome Location:
Chr.20: 30578966 - 30610172 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03070972_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cep43
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002125.1 NP_001002125.1 5 - 6 438 75
XM_005160662.4 XP_005160719.1 5 - 6 472 75
XM_005160663.4 XP_005160720.1 5 - 6 472 75
XM_005160664.4 XP_005160721.1 5 - 6 472 75
XM_017352461.2 XP_017207950.1 5 - 6 472 75
XM_021468390.1 XP_021324065.1 5 - 6 472 75
XM_021468391.1 XP_021324066.1 5 - 6 472 75
XM_021468392.1 XP_021324067.1 5 - 6 472 75
XM_021468393.1 XP_021324068.1 5 - 6 472 75
GenBank mRNA BC071450.1 - 5 - 6 438 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

microtubule anchoring
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0