This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02370171_s1
See other MT2 GE Assays ›
Gene Symbol:
Mt2
Gene Name:
Methyltransferase 2
Gene Aliases:
CG10692, DNMT, DNMT2, DmMT2, Dmel\CG10692, Dmel_CG10692, Dnmt, Dnmt2, dDNMT, dDNMT2, dDnmt2, dnmt2, nmt2
Chromosome Location:
Chr.2L: 12096525 - 12097687 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0028707
Assay ID Dm02370171_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mt2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042890.2 NP_001036355.1 2 510 65
NM_058127.4 NP_477475.2 2 485 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster Inventoried

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

telomere maintenance
positive regulation of defense response to virus by host
DNA methylation
response to oxidative stress
determination of adult lifespan
response to heat
gene silencing
tRNA methylation
regulation of tRNA stability
DNA methylation involved in embryo development
positive regulation of innate immune response
tRNA binding
RNA binding
DNA (cytosine-5-)-methyltransferase activity
methyltransferase activity
tRNA methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0