This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02368780_g1
See other EMC2A GE Assays ›
Gene Symbol:
EMC2A EMC2B
Gene Name:
ER membrane protein complex subunit 2A
ER membrane protein complex subunit 2B
Gene Aliases:
CG17556, Dmel\CG17556, Dmel_CG17556, emc2A
CG3678, Dmel\CG3678, Dmel_CG3678, RE62710p, emc2B
Chromosome Location:
Chr.3R: 16994542 - 16996071 on Build Release_6_plus_ISO1_MT
Chromosome Location:
Chr.3R: 16990334 - 16991878 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0038462
FBgn0038461
Assay ID Dm02368780_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EMC2A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_142324.2 NP_650581.1 4 1038 135
GenBank mRNA BT100172.1 - 4 135
Gene Symbol: EMC2B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001275722.1 NP_001262651.1 4 135
GenBank mRNA BT120319.1 - 4 993 135

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0