This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02361968_s1
See other BEN GE Assays ›
Gene Symbol:
ben
Gene Name:
bendless
Gene Aliases:
BEN, Ben, Bend, CG18319, Dmel\CG18319, Dmel_CG18319, UBC13, Ubc13, UbcD3, anon-WO03040301.242, bend, nj-262, ubc13
Chromosome Location:
Chr.X: 13996345 - 13999805 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0000173
Assay ID Dm02361968_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ben
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001258734.2 NP_001245663.1 3 1541 107
NM_001272611.2 NP_001259540.1 2 1390 107
NM_001272612.2 NP_001259541.1 2 1348 107
NM_078595.3 NP_511150.1 2 1428 107
GenBank mRNA S70118.1 - 2 1367 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
postreplication repair
response to oxidative stress
axonogenesis
axon target recognition
grooming behavior
flight behavior
jump response
photoreceptor cell morphogenesis
positive regulation of cell death
positive regulation of mitotic cell cycle, embryonic
positive regulation of JNK cascade
synaptic growth at neuromuscular junction
synapse maturation
peptidoglycan recognition protein signaling pathway
protein K63-linked ubiquitination
response to anesthetic
positive regulation of protein polyubiquitination
ubiquitin conjugating enzyme binding
ubiquitin protein ligase binding
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0