This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02361909_s1
See other ACT5C GE Assays ›
Gene Symbol:
Act5C
Gene Name:
Actin 5C
Gene Aliases:
A, A4V404_DROME, ACT, ACT1_DROME, ACT5C, Ac5C, Act, Act-5C, Act5, Act5c, Actin, Actin/BAP47, Actin5C, ArpA, BAP47, Bap47, CG4027, Dmel\CG4027, Dmel_CG4027, F-actin, G-actin, M32055, T11, act, act 5C, act5C, actin, actin5C, anon-EST:fe2D2, beta-actin, beta-actin/Bap47, chrX:5748184..5748304, cyt5C, l(1)G0009, l(1)G0010, l(1)G0025, l(1)G0079, l(1)G0117, l(1)G0177, l(1)G0245, l(1)G0330, l(1)G0420, l(1)G0486
Chromosome Location:
Chr.X: 5900861 - 5905399 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0000042
Assay ID Dm02361909_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Act5C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001014725.2 NP_001014725.1 2 1475 92
NM_001014726.2 NP_001014726.1 2 1475 92
NM_001297986.1 NP_001284915.1 2 1475 92
NM_078497.4 NP_511052.1 2 1437 92
NM_167053.2 NP_727048.1 2 1475 92
GenBank mRNA AY089562.1 - 2 1469 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster Inventoried

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

Gene Ontology Categories:

Function(s)

mitotic cytokinesis
chromatin remodeling
sperm individualization
maintenance of protein location in cell
tube formation

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0