This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02332442_g1
See other DPY GE Assays ›
Gene Symbol:
dpy
Gene Name:
dumpy
Gene Aliases:
BcDNA:SD02173, CG15637, CG31724, CG31725, CG31726, CG31727, CG31964, CG31965, CG31966, CG31967, CG31968, CG33000, CG33086, CG33196, CR33195, CT35799, DP, DPY, Dmel\CG33196, Dmel_CG33196, Dp, Dpy, Mur24F, Q8IQ18_DROME, SP460, dp, l(2)24Fd, olv[D]
Chromosome Location:
Chr.2L: 4477462 - 4614300 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0053196
Assay ID Dm02332442_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dpy
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001258946.1 NP_001245875.1 24 - 25 29828 141
NM_001258947.3 NP_001245876.1 26 - 27 30046 141
NM_001258950.1 NP_001245879.1 17 - 18 8651 141
NM_001273101.1 NP_001260030.1 24 - 25 29200 141
NM_001273102.1 NP_001260031.1 24 - 25 29828 141
NM_001273103.1 NP_001260032.1 24 - 25 29828 141
NM_001273104.1 NP_001260033.1 23 - 24 29471 141
NM_001273105.1 NP_001260034.1 20 - 21 16904 141
NM_001273106.1 NP_001260035.1 24 - 25 29828 141
NM_001273107.1 NP_001260036.1 24 - 25 29828 141
NM_001273108.1 NP_001260037.1 22 - 23 22193 141
NM_001273109.1 NP_001260038.1 23 - 24 28978 141
NM_001273110.1 NP_001260039.1 23 - 24 28978 141
NM_001273111.1 NP_001260040.1 18 - 19 15256 141
NM_001273112.1 NP_001260041.1 17 - 18 15266 141
NM_001273113.1 NP_001260042.1 24 - 25 29828 141

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
open tracheal system development
apposition of dorsal and ventral imaginal disc-derived wing surfaces
chitin-based embryonic cuticle biosynthetic process
chitin-based cuticle attachment to epithelium
regulation of imaginal disc-derived wing size
transcription factor activity, sequence-specific DNA binding
extracellular matrix structural constituent
calcium ion binding
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0