This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02152402_g1
See other ACF GE Assays ›
Gene Symbol:
Acf
Gene Name:
ATP-dependent chromatin assembly factor large subunit
Gene Aliases:
175K protein, ACF, ACF/CHRAC, ACF1, Acf-1, Acf1, CAF, CG1966, CHRAC, CHRAC-175, Dmel\CG1966, Dmel_CG1966, acf1, dACF, dCHRAC, p170, p170/p185, p175
Chromosome Location:
Chr.3R: 31793981 - 31799799 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0027620
Assay ID Dm02152402_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080486.3 NP_536734.2 4 - 5 4274 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

chromatin assembly or disassembly
nucleosome assembly
regulation of transcription, DNA-templated
nervous system development
nucleosome positioning
regulation of heterochromatin assembly
chromatin assembly
nucleosome mobilization
regulation of ATPase activity
negative regulation of transcription, DNA-templated
DNA binding
histone acetyltransferase activity
protein binding
nucleosome-dependent ATPase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header