This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm02150796_g1
See other SLO GE Assays ›
Gene Symbol:
slo
Gene Name:
slowpoke
Gene Aliases:
BcDNA:GH10751, CG10693, Dmel\CG10693, Dmel_CG10693, Dslo, Slo, Slo1, dSlo, dSlo1, dslo, fSlo
Chromosome Location:
Chr.3R: 24662491 - 24711595 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0003429
Assay ID
Dm02150796_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: slo
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001014651.3
NP_001014651.1
3 - 4
1266
174
NM_001014652.3
NP_001014652.1
3 - 4
1266
174
NM_001014653.3
NP_001014653.1
3 - 4
1266
174
NM_001014654.3
NP_001014654.1
3 - 4
1266
174
NM_001014655.3
NP_001014655.1
3 - 4
1266
174
NM_001014656.3
NP_001014656.1
3 - 4
1266
174
NM_001014657.3
NP_001014657.1
3 - 4
1266
174
NM_001014658.3
NP_001014658.1
3 - 4
1266
174
NM_001014659.3
NP_001014659.1
3 - 4
1266
174
NM_001014660.3
NP_001014660.1
3 - 4
1266
174
NM_001014661.4
NP_001014661.2
4 - 5
1007
174
NM_001014662.3
NP_001014662.1
3 - 4
1266
174
NM_001014663.3
NP_001014663.1
3 - 4
1266
174
NM_001014664.3
NP_001014664.1
3 - 4
1266
174
NM_001170241.3
NP_001163712.1
5 - 6
1535
174
NM_001275995.1
NP_001262924.1
5 - 6
1196
174
NM_001275996.1
NP_001262925.1
3 - 4
1266
174
NM_001275997.1
NP_001262926.1
4 - 5
1007
174
NM_001275998.1
NP_001262927.1
4 - 5
1007
174
NM_001275999.1
NP_001262928.1
4 - 5
1007
174
NM_079762.4
NP_524486.2
3 - 4
1266
174
NM_170164.4
NP_733029.1
4 - 5
1007
174
NM_206566.4
NP_996289.1
4 - 5
1007
174
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Drosophila melanogaster
Panther Classification:
Molecular Function -
ion channel
Gene Ontology Categories:
Function(s)
Process(es)
potassium ion transport
circadian rhythm
regulation of synaptic growth at neuromuscular junction
response to drug
male courtship behavior, veined wing generated song production
circadian behavior
transmembrane transport
negative regulation of neuromuscular synaptic transmission
protein binding
calcium-activated potassium channel activity
large conductance calcium-activated potassium channel activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header