This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02150795_m1
See other SLO GE Assays ›
Gene Symbol:
slo
Gene Name:
slowpoke
Gene Aliases:
BcDNA:GH10751, CG10693, Dmel\CG10693, Dmel_CG10693, Dslo, Slo, Slo1, dSlo, dSlo1, dslo, fSlo
Chromosome Location:
Chr.3R: 24662491 - 24711595 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0003429
Assay ID Dm02150795_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slo
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001014651.3 NP_001014651.1 2 - 3 1102 104
NM_001014652.3 NP_001014652.1 2 - 3 1102 104
NM_001014653.3 NP_001014653.1 2 - 3 1102 104
NM_001014654.3 NP_001014654.1 2 - 3 1102 104
NM_001014655.3 NP_001014655.1 2 - 3 1102 104
NM_001014656.3 NP_001014656.1 2 - 3 1102 104
NM_001014657.3 NP_001014657.1 2 - 3 1102 104
NM_001014658.3 NP_001014658.1 2 - 3 1102 104
NM_001014659.3 NP_001014659.1 2 - 3 1102 104
NM_001014660.3 NP_001014660.1 2 - 3 1102 104
NM_001014661.4 NP_001014661.2 3 - 4 843 104
NM_001014662.3 NP_001014662.1 2 - 3 1102 104
NM_001014663.3 NP_001014663.1 2 - 3 1102 104
NM_001014664.3 NP_001014664.1 2 - 3 1102 104
NM_001170241.3 NP_001163712.1 4 - 5 1371 104
NM_001275995.1 NP_001262924.1 4 - 5 1032 104
NM_001275996.1 NP_001262925.1 2 - 3 1102 104
NM_001275997.1 NP_001262926.1 3 - 4 843 104
NM_001275998.1 NP_001262927.1 3 - 4 843 104
NM_001275999.1 NP_001262928.1 3 - 4 843 104
NM_079762.4 NP_524486.2 2 - 3 1102 104
NM_170164.4 NP_733029.1 3 - 4 843 104
NM_206566.4 NP_996289.1 3 - 4 843 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Drosophila melanogaster Inventoried

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

potassium ion transport
circadian rhythm
regulation of synaptic growth at neuromuscular junction
response to drug
male courtship behavior, veined wing generated song production
circadian behavior
transmembrane transport
negative regulation of neuromuscular synaptic transmission
protein binding
calcium-activated potassium channel activity
large conductance calcium-activated potassium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0