This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02150783_g1
See other SLO GE Assays ›
Gene Symbol:
slo
Gene Name:
slowpoke
Gene Aliases:
BcDNA:GH10751, CG10693, Dmel\CG10693, Dmel_CG10693, Dslo, Slo, Slo1, dSlo, dSlo1, dslo, fSlo
Chromosome Location:
Chr.3R: 24662491 - 24711595 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0003429
Assay ID Dm02150783_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slo
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001014651.3 NP_001014651.1 13 - 14 2585 70
NM_001014652.3 NP_001014652.1 13 - 14 2585 70
NM_001014653.3 NP_001014653.1 13 - 14 2585 70
NM_001014654.3 NP_001014654.1 13 - 14 2585 70
NM_001014655.3 NP_001014655.1 13 - 14 2585 70
NM_001014656.3 NP_001014656.1 13 - 14 2585 70
NM_001014657.3 NP_001014657.1 13 - 14 2585 70
NM_001014658.3 NP_001014658.1 13 - 14 2585 70
NM_001014659.3 NP_001014659.1 13 - 14 2585 70
NM_001014660.3 NP_001014660.1 13 - 14 2585 70
NM_001014661.4 NP_001014661.2 14 - 15 2326 70
NM_001014662.3 NP_001014662.1 13 - 14 2585 70
NM_001014663.3 NP_001014663.1 13 - 14 2585 70
NM_001014664.3 NP_001014664.1 13 - 14 2585 70
NM_001170241.3 NP_001163712.1 15 - 16 2854 70
NM_001275995.1 NP_001262924.1 15 - 16 2515 70
NM_001275996.1 NP_001262925.1 13 - 14 2585 70
NM_001275997.1 NP_001262926.1 14 - 15 2326 70
NM_001275998.1 NP_001262927.1 14 - 15 2326 70
NM_001275999.1 NP_001262928.1 14 - 15 2326 70
NM_079762.4 NP_524486.2 13 - 14 2585 70
NM_170164.4 NP_733029.1 14 - 15 2326 70
NM_206566.4 NP_996289.1 14 - 15 2326 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

potassium ion transport
circadian rhythm
regulation of synaptic growth at neuromuscular junction
response to drug
male courtship behavior, veined wing generated song production
circadian behavior
transmembrane transport
negative regulation of neuromuscular synaptic transmission
protein binding
calcium-activated potassium channel activity
large conductance calcium-activated potassium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0