This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02149531_g1
See other MHCL GE Assays ›
Gene Symbol:
Mhcl
Gene Name:
Myosin heavy chain-like
Gene Aliases:
CG10218, CG10224, CG31045, DmPDZ, Dmel\CG31045, Dmel_CG31045, MHCL(PDZ), Myo7A
Chromosome Location:
Chr.3R: 16068523 - 16098761 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0026059
Assay ID Dm02149531_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mhcl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001043250.2 NP_001036715.1 8 - 9 3270 86
NM_001043251.2 NP_001036716.1 5 - 6 2997 86
NM_169700.3 NP_732108.2 8 - 9 3270 86
NM_169701.3 NP_732109.2 8 - 9 3270 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

biological_process
microfilament motor activity
ATP binding
actin-dependent ATPase activity
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0