This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02148065_g1
See other SEC23 GE Assays ›
Gene Symbol:
Sec23
Gene Name:
Secretory 23
Gene Aliases:
CG1250, COPII, Dmel\CG1250, Dmel_CG1250, Hau, SEC23, Sec23A, Sec23p, dSec23, dSec23p, dar2, dsec23, dsec23p, hau, sec23
Chromosome Location:
Chr.3R: 5649065 - 5652629 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0262125
Assay ID Dm02148065_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sec23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001275365.1 NP_001262294.1 5 - 6 2341 77
NM_001300255.1 NP_001287184.1 5 - 6 2428 77
NM_169088.2 NP_730978.1 5 - 6 2457 77
NM_169089.4 NP_730979.1 4 - 5 2549 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Drosophila melanogaster

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of extracellular matrix constituent secretion
regulation of COPII vesicle coating
intracellular protein transport
endoplasmic reticulum organization
Golgi organization
regulation of cell shape
larval chitin-based cuticle development
muscle attachment
positive regulation of cell-cell adhesion
maintenance of cell polarity
protein localization to cell surface
lumen formation, open tracheal system
chitin-based larval cuticle pattern formation
chitin-based cuticle development
positive regulation of GTPase activity
positive regulation of cuticle pigmentation
positive regulation of dendrite morphogenesis
basement membrane organization
protein localization to plasma membrane
GTPase activator activity
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0