This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02147941_g1
See other ITPR GE Assays ›
Gene Symbol:
Itpr
Gene Name:
Inositol 1,4,5,-trisphosphate receptor
Gene Aliases:
CG1063, DIP, DmInsP[[3R]], DmInsP[[3]]R, Dmel\CG1063, Dmel_CG1063, IP, IP3R, IP[[3]] receptor, IP[[3]]R, ITP-r83A, ITPR1, Ins3PR, InsP(3), InsP3, InsP3R, InsP[[3]], InsP[[3]]R, Insp3R, Ip3R, Itp-R83A, Itp-r, Itp-r-83A, Itp-r42B, Itp-r83A, Itp-r[1], anon-WO02059370.49, dIP3R, dIP[[3]]R, dip, itpr, itpr-83, l(3)05616, l(3)itpr, l(3)j5B4
Chromosome Location:
Chr.3R: 5517117 - 5539411 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0010051
Assay ID Dm02147941_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Itpr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001300251.1 NP_001287180.1 18 - 19 8974 73
NM_169060.2 NP_730941.1 19 - 20 8956 73
NM_169061.2 NP_730942.1 18 - 19 8929 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster Inventoried

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

nuclear division
calcium ion transport
cellular calcium ion homeostasis
response to oxidative stress
molting cycle, chitin-based cuticle
phototransduction
flight behavior
cellular response to starvation
negative regulation of lipid storage
mushroom body development
stabilization of membrane potential
larval feeding behavior
salivary gland cell autophagic cell death
response to starvation
positive regulation of ecdysteroid secretion
sensory perception of taste
release of sequestered calcium ion into cytosol
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
lipid homeostasis
fatty acid homeostasis
regulation of feeding behavior
calcium ion transmembrane transport
cellular response to ethanol
inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
calcium ion binding
phosphatidylinositol binding
inositol 1,4,5 trisphosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0