This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02140513_g1
See other SDR GE Assays ›
Gene Symbol:
Sdr
Gene Name:
Secreted decoy of InR
Gene Aliases:
CG3837, Dmel\CG3837, Dmel_CG3837, SDR
Chromosome Location:
Chr.3R: 14918628 - 14923083 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0038279
Assay ID Dm02140513_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sdr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001300402.1 NP_001287331.1 6 - 7 2362 63
NM_142151.2 NP_650408.1 6 - 7 2362 63
GenBank mRNA BT133291.1 - 6 - 7 2311 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Drosophila melanogaster

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
negative regulation of multicellular organism growth
response to starvation
negative regulation of insulin receptor signaling pathway
transmembrane receptor protein tyrosine kinase activity
insulin-like growth factor binding
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0