Assay Details
Gene Symbol: Pif1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170085.2 | NP_001163556.2 | 2 | 1089 | 69 | |
NM_001202266.2 | NP_001189195.2 | 2 | 908 | 69 | ||
NM_001316487.1 | NP_001303416.1 | 3 | 994 | 69 |
Gene Symbol: Pif1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275457.3 | NP_001262386.1 | 2 | 1089 | 69 | |
NM_001275458.2 | NP_001262387.1 | 3 | 994 | 69 | ||
NM_001316570.1 | NP_001303499.1 | 2 | 908 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001275457.3, NM_001275458.2, NM_001316570.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
