This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02137032_m1
See other PEK GE Assays ›
Gene Symbol:
PEK
Gene Name:
pancreatic eIF-2alpha kinase
Gene Aliases:
CG2087, DPERK, DmPEK, Dmel\CG2087, Dmel_CG2087, DpERK, PERK, Pek, Perk, dPERK, dPerk, pERK, pek, perk
Chromosome Location:
Chr.3R: 5458563 - 5464753 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0037327
Assay ID Dm02137032_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PEK
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_141281.3 NP_649538.1 1 - 2 532 75
GenBank mRNA AJ313085.1 - 1 - 2 492 75
AY094831.1 - 1 - 2 508 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Drosophila melanogaster

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

regulation of G2/M transition of mitotic cell cycle
positive regulation of autophagy
PERK-mediated unfolded protein response
positive regulation of JNK cascade
protein kinase activity
elongation factor-2 kinase activity
eukaryotic translation initiation factor 2alpha kinase activity
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header