This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02136824_g1
See other SPARTIN GE Assays ›
Gene Symbol:
spartin
Gene Name:
CG12001-PA
Gene Aliases:
CG12001, Dmel\CG12001, Dmel_CG12001, Spartin, dSp
Chromosome Location:
Chr.3R: 4654800 - 4657985 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0037265
Assay ID Dm02136824_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: spartin
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001259981.1 NP_001246910.1 3 - 4 1559 73
NM_141228.2 NP_649485.1 3 - 4 1727 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of receptor internalization
negative regulation of BMP signaling pathway
negative regulation of synaptic growth at neuromuscular junction
cell division
Rab GTPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header