This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02136186_g1
See other CORTACTIN GE Assays ›
Gene Symbol:
Cortactin
Gene Name:
CG3637-PA
Gene Aliases:
CG3637, Dmel\CG3637, Dmel_CG3637, cortactin, cttn
Chromosome Location:
Chr.3R: 21061680 - 21064471 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0025865
Assay ID Dm02136186_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cortactin
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001260293.2 NP_001247222.1 3 - 4 765 59
NM_001260294.1 NP_001247223.1 4 - 5 737 59
NM_001260295.1 NP_001247224.1 4 - 5 734 59
NM_079702.3 NP_524426.2 3 - 4 762 59
GenBank mRNA AY051774.1 - 3 - 4 710 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster Inventoried

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein cytoskeletal protein non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

border follicle cell migration
female germline ring canal formation
regulation of actin filament polymerization
positive regulation of receptor-mediated endocytosis
actin filament binding
proline-rich region binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0