This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02135840_g1
See other SU(VAR)3-9 GE Assays ›
Gene Symbol:
Su(var)3-9 eIF2gamma
Gene Name:
Suppressor of variegation 3-9
eukaryotic translation initiation factor 2 subunit gamma
Gene Aliases:
CG43664, CG6476, Dmel\CG43664, Dmel_CG43664, E(var)3-2, SU(VAR)3-9, SUV39, SUV39H1, Su(VAR)3-9, Su(Var)3-9, Su(var), Su(var) 3-9, Su(var)3-902, Su(var)306, Su(var)310, Su(var)328, Su-var(3)9, Su-var(3)902, SuVar3-9, Suv 3-9, Suvar(3)9, Suvar3-9, Suvar39, caa56376 Suvar39 Dm, dSU(VAR)3-9, dSu(var)3-9, ptn, su(var)3-9
51kDa protein, CG43665, CG6476, Dmel\CG43665, Dmel_CG43665, eIF-2G, eIF-2gamma, eIF2G
Chromosome Location:
Chr.3R: 15259864 - 15263458 on Build Release_6_plus_ISO1_MT
Chromosome Location:
Chr.3R: 15257844 - 15263458 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0263755
FBgn0263740
Assay ID Dm02135840_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Su(var)3-9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_079633.3 NP_524357.2 1 - 2 179 70
GenBank mRNA BT133458.1 - 1 - 2 159 70
Gene Symbol: eIF2gamma
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001275658.1 NP_001262587.1 1 - 2 179 70
NM_169629.2 NP_731993.1 1 - 2 179 70
GenBank mRNA AY061102.1 - 1 - 2 137 70
X80069.1 - 157 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein histone modifying enzyme translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

DNA methylation
chromatin organization
chromatin silencing
chromatin silencing at telomere
gene silencing
histone modification
histone methylation
heterochromatin assembly
pericentric heterochromatin assembly
histone lysine methylation
female germ-line stem cell asymmetric division
oogenesis
chromosome organization
histone H3-K9 methylation
heterochromatin organization
heterochromatin organization involved in chromatin silencing
negative regulation of response to gamma radiation
formation of translation preinitiation complex
positive regulation of translational fidelity
chromatin binding
zinc ion binding
histone methyltransferase activity
histone methyltransferase activity (H3-K9 specific)
tRNA binding
translation initiation factor activity
GTPase activity
GTP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0