This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02135789_m1
See other SIM GE Assays ›
Gene Symbol:
sim
Gene Name:
single-minded
Gene Aliases:
0483/09, 0716/08, 0899/14, 0953/08, 1002/10, 1034/02, 1050/13, 1110/04, 1111/10, 1330/08, 1479/04, CG7771, Dmel\CG7771, Dmel_CG7771, S8, SIM, Sim, bHLHe16, l(3)87Ea, l(3)E320, l(3)RD, l(3)S8, l(3)s8, schm, shm
Chromosome Location:
Chr.3R: 13057755 - 13078124 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0004666
Assay ID Dm02135789_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: sim
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001275598.2 NP_001262527.1 6 - 7 1047 70
NM_001275599.1 NP_001262528.1 6 - 7 1351 70
NM_079616.4 NP_524340.2 6 - 7 990 70
NM_169495.4 NP_731771.3 6 - 7 1327 70
GenBank mRNA AY129457.1 - 6 - 7 987 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
ectoderm development
axonogenesis
axon guidance
ventral midline development
ventral cord development
brain development
adult walking behavior
determination of genital disc primordium
locomotion
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0