This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01845369_g1
See other NAA20A GE Assays ›
Gene Symbol:
Naa20A
Gene Name:
N(alpha)-acetyltransferase 20 A
Gene Aliases:
CG14222, DmAAF48987, Dmel\CG14222, Dmel_CG14222, NAA20, Naa20, dNAA20
Chromosome Location:
Chr.X: 19621451 - 19623074 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0031043
Assay ID Dm01845369_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Naa20A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134487.3 NP_608331.1 5 - 6 1026 68
GenBank mRNA AY061383.1 - 5 - 6 964 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Drosophila melanogaster

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

histone acetylation
N-terminal peptidyl-methionine acetylation
negative regulation of neuron apoptotic process
peptide alpha-N-acetyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0