This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01843624_g1
See other RBF GE Assays ›
Gene Symbol:
Rbf
Gene Name:
Retinoblastoma-family protein
Gene Aliases:
CG7413, Dmel\CG7413, Dmel_CG7413, EG:34F3.3, FBF, RB, RBF, RBF1, RBf1, Rb, Rb1, RbF, Rbf1, dRBF, pRb, rb, rbf, rbf1
Chromosome Location:
Chr.X: 797090 - 801479 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0015799
Assay ID Dm01843624_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080297.3 NP_525036.2 7 - 8 3018 73
GenBank mRNA AY089532.1 - 7 - 8 2996 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Drosophila melanogaster

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
endomitotic cell cycle
eggshell chorion gene amplification
regulation of mitotic cell cycle
negative regulation of DNA replication
regulation of cell size
intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of gene expression
regulation of cell death
cell differentiation
maintenance of mitotic sister chromatid cohesion
DNA endoreduplication
regulation of cell proliferation
response to starvation
negative regulation of apoptotic process
regulation of DNA binding
regulation of chromosome segregation
regulation of cohesin loading
regulation of execution phase of apoptosis
negative regulation of G1/S transition of mitotic cell cycle
transcription regulatory region sequence-specific DNA binding
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II activating transcription factor binding
protein binding
transcription factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0