This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01843079_m1
See other CV-2 GE Assays ›
Gene Symbol:
cv-2
Gene Name:
crossveinless 2
Gene Aliases:
BMPER, CG15671, CT35855, CV2, Cv 2, Cv-2, Cv2, Dmel\CG15671, Dmel_CG15671, cv 2
Chromosome Location:
Chr.2R: 21355015 - 21379464 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0000395
Assay ID Dm01843079_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cv-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080070.4 NP_524809.2 3 - 4 1351 73
GenBank mRNA BT133425.1 - 3 - 4 1349 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

imaginal disc-derived wing vein specification
imaginal disc-derived wing morphogenesis
positive regulation of BMP signaling pathway
BMP binding
heparan sulfate proteoglycan binding
BMP receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0