This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01842879_g1
See other RACK1 GE Assays ›
Gene Symbol:
Rack1
Gene Name:
Receptor of activated protein kinase C 1
Gene Aliases:
CG7111, Dmel\CG7111, Dmel_CG7111, RACK1, dRack1, i173, rack1
Chromosome Location:
Chr.2L: 7825347 - 7827315 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0020618
Assay ID Dm01842879_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rack1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001273288.1 NP_001260217.1 1 - 2 189 63
NM_001273289.1 NP_001260218.1 1 - 2 189 63
NM_001298794.1 NP_001285723.1 1 - 2 258 63
NM_057921.4 NP_477269.1 1 - 2 189 63
GenBank mRNA AY071661.1 - 1 - 2 222 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
locomotory behavior
cellular response to starvation
regulation of autophagosome size
oviposition
positive regulation of actin filament polymerization
wing disc development
cuticle development
positive regulation by host of viral genome replication
positive regulation of glycogen biosynthetic process
muscle cell cellular homeostasis
oogenesis
rescue of stalled ribosome
IRES-dependent viral translational initiation
protein kinase C binding
enzyme binding
ribosome binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0