This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01842798_g1
See other CDK9 GE Assays ›
Gene Symbol:
Cdk9
Gene Name:
Cyclin-dependent kinase 9
Gene Aliases:
CDK9, CG5179, DmP-TEFb, Dmel\CG5179, Dmel_CG5179, P-TEF, P-TEFb, PTefb, cdk9
Chromosome Location:
Chr.2R: 22636063 - 22637727 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0019949
Assay ID Dm01842798_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdk9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_057878.4 NP_477226.2 2 - 3 405 69
GenBank mRNA AF027300.1 - 2 - 3 382 69
AY118809.1 - 2 - 3 387 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
transcription, DNA-templated
transcription elongation from RNA polymerase II promoter
protein phosphorylation
response to heat
positive regulation of transcription from RNA polymerase II promoter
cyclin-dependent protein serine/threonine kinase activity
ATP binding
RNA polymerase II carboxy-terminal domain kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0