This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01841325_g1
See other LSP1BETA GE Assays ›
Gene Symbol:
Lsp1beta
Gene Name:
Larval serum protein 1 beta
Gene Aliases:
CG4178, DmeLSP1b, Dmel\CG4178, Dmel_CG4178, LSP 1, LSP-1, LSP-1 beta, LSP-1beta, LSP1, LSP1beta, Lsp-1b, Lsp-1beta, Lsp1, Lsp1 beta, Lsp1-beta, Lsp1[beta], Lsp1b
Chromosome Location:
Chr.2L: 898500 - 901316 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0002563
Assay ID Dm01841325_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lsp1beta
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001298627.1 NP_001285556.1 1 - 2 437 82
NM_057276.4 NP_476624.1 1 - 2 293 82
GenBank mRNA AY069163.1 - 1 - 2 271 82
BT024223.1 - 2841 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0