This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01840909_m1
See other CERT GE Assays ›
Gene Symbol:
cert
Gene Name:
ceramide transfer protein
Gene Aliases:
BcDNA:GH07688, CERT, CG7207, Dcert, Dmel\CG7207, Dmel_CG7207, dcert
Chromosome Location:
Chr.3L: 8282528 - 8289870 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0027569
Assay ID Dm01840909_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cert
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001300049.1 NP_001286978.1 4 - 5 913 109
NM_139942.5 NP_648199.1 5 - 6 1076 109
GenBank mRNA AF145644.1 - 5 - 6 1037 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Drosophila melanogaster

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

sphingolipid metabolic process
ER to Golgi ceramide transport
ceramide transport
intermembrane lipid transfer
lipid binding
phosphatidylinositol-4-phosphate binding
ceramide binding
intermembrane ceramide transfer activity
ceramide 1-phosphate binding
ceramide 1-phosphate transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0