This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01840471_g1
See other EIF4E4 GE Assays ›
Gene Symbol:
eIF4E4
Gene Name:
eukaryotic translation initiation factor 4E4
Gene Aliases:
BEST:GH01027, CG10124, CT28485, Dmel\CG10124, Dmel_CG10124, NEST:bs04e06, eIF4E, eIF4E-4
Chromosome Location:
Chr.3L: 6664224 - 6665641 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0035709
Assay ID Dm01840471_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eIF4E4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_139795.3 NP_648052.1 2 - 3 576 67
GenBank mRNA BT021432.1 - 2 - 3 568 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster Inventoried

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
RNA cap binding
RNA 7-methylguanosine cap binding
translation initiation factor activity
protein binding
eukaryotic initiation factor 4G binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0