This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01834235_g1
See other USP15-31 GE Assays ›
Gene Symbol:
Usp15-31
Gene Name:
Ubiquitin specific protease 15/31
Gene Aliases:
CG30421, CG3872, CG9189, DUSP31, Dmel\CG30421, Dmel_CG30421, Q9W117, USP43, Usp15, Usp31
Chromosome Location:
Chr.2R: 24743749 - 24756976 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0050421
Assay ID Dm01834235_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Usp15-31
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001299926.1 NP_001286855.1 4 - 5 3419 108
NM_001299927.1 NP_001286856.1 4 - 5 3419 108
NM_138115.3 NP_611959.2 5 - 6 3503 108
GenBank mRNA BT057999.1 - 5 - 6 3440 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Drosophila melanogaster

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
protein deubiquitination
thiol-dependent ubiquitin-specific protease activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0