This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01831627_m1
See other EIF5B GE Assays ›
Gene Symbol:
eIF5B
Gene Name:
eukaryotic translation initiation factor 5B
Gene Aliases:
A1, CG10840, CG14970, Dmel\CG10840, Dmel_CG10840, Eif5b, IF2, anon-WO0118547.362, cIF2, dIF2, yIF2
Chromosome Location:
Chr.3L: 3428658 - 3460977 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0026259
Assay ID Dm01831627_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eIF5B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001259670.2 NP_001246599.1 5 - 6 3414 59
NM_001259671.2 NP_001246600.1 4 - 5 3143 59
NM_001259672.2 NP_001246601.1 4 - 5 3143 59
NM_001259673.2 NP_001246602.1 5 - 6 3439 59
NM_143717.3 NP_651974.1 4 - 5 3143 59
GenBank mRNA BT150280.1 - 4 - 5 3142 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster Inventoried

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
translation initiation factor activity
GTPase activity
protein binding
GTP binding
DEAD/H-box RNA helicase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0