This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01830721_g1
See other ICE1 GE Assays ›
Gene Symbol:
Ice1
Gene Name:
Interacts with the C terminus of ELL 1
Gene Aliases:
CG13550, Dmel\CG13550, Dmel_CG13550, ICE1, dICE1
Chromosome Location:
Chr.2R: 23366866 - 23371459 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0034853
Assay ID Dm01830721_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ice1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_137942.2 NP_611786.1 1 - 2 1559 68
GenBank mRNA BT150380.1 - 1 - 2 1536 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

snRNA transcription from RNA polymerase II promoter
chromatin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0