This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01828675_g1
See other RHOGAPP190 GE Assays ›
Gene Symbol:
RhoGAPp190
Gene Name:
Rho GTPase activating protein p190
Gene Aliases:
8267, BcDNA:GH17919, BcDNA:GM05133, CG32555, CG8240, CG8267, D-p190, DRhoGAP, Dmel\CG32555, Dmel_CG32555, RhoGAP, RhoGAP P190, RhoGAP-16B12, mdcds_56566, p190, p190 RhoGAP, p190-RhoGAP, p190RhoGAP
Chromosome Location:
Chr.X: 17639147 - 17652562 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0026375
Assay ID Dm01828675_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RhoGAPp190
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001272727.1 NP_001259656.1 5 - 6 2858 82
NM_001272728.1 NP_001259657.1 5 - 6 3205 82
NM_001272729.1 NP_001259658.1 5 - 6 2797 82
NM_001272730.1 NP_001259659.1 5 - 6 2797 82
NM_133003.3 NP_573231.2 5 - 6 2797 82
NM_167572.2 NP_728089.1 5 - 6 2914 82
NM_167573.2 NP_728090.1 5 - 6 3205 82
GenBank mRNA AF387518.1 - 4 - 5 2723 82
BT150459.1 - 5 - 6 2787 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Drosophila melanogaster

Panther Classification:

Molecular Function -

G-protein modulator protein-binding activity modulator GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

Rho protein signal transduction
defasciculation of motor neuron axon
mushroom body development
negative regulation of cell size
regulation of axonogenesis
GTPase activity
GTPase activator activity
GTP binding
semaphorin receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0