This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01828117_g1
See other UP GE Assays ›
Gene Symbol:
up
Gene Name:
upheld
Gene Aliases:
BcDNA:LD08591, CG7107, DmTpnT, Dmel\CG7107, Dmel_CG7107, TNT, Tn-T, TnT, TpnT, cTNT, dro TnT, int, tnT, upheld2, wupB
Chromosome Location:
Chr.X: 13593474 - 13602578 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0004169
Assay ID
Dm01828117_g1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
374.00
Your Price
Online offer:
374.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: up
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001014737.2
NP_001014737.1
7 - 8
894
62
NM_001014738.3
NP_001014738.2
7 - 8
894
62
NM_001014739.2
NP_001014739.1
5 - 6
828
62
NM_001169270.2
NP_001162741.1
5 - 6
828
62
NM_001169271.2
NP_001162742.1
7 - 8
897
62
NM_001169272.2
NP_001162743.1
6 - 7
873
62
NM_001169273.2
NP_001162744.1
6 - 7
942
62
NM_001258723.2
NP_001245652.1
5 - 6
825
62
NM_001258724.2
NP_001245653.1
7 - 8
898
62
NM_001272593.2
NP_001259522.1
7 - 8
969
62
NM_001272594.2
NP_001259523.1
7 - 8
894
62
NM_001272595.2
NP_001259524.1
6 - 7
870
62
NM_001298284.1
NP_001285213.1
6 - 7
918
62
NM_001298285.1
NP_001285214.1
6 - 7
1068
62
NM_001298286.1
NP_001285215.1
4 - 5
785
62
NM_080349.3
NP_525088.2
7 - 8
897
62
NM_167375.2
NP_727718.1
6 - 7
945
62
NM_167376.2
NP_727719.1
4 - 5
1056
62
GenBank mRNA
AY051989.1
-
4 - 5
1056
62
AY665838.1
-
4 - 5
714
62
BT058030.1
-
5 - 6
797
62
BT125908.1
-
7 - 8
873
62
BT126178.1
-
6 - 7
864
62
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
3' Most
Recommended Assay
Drosophila melanogaster
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
cellular calcium ion homeostasis
muscle contraction
regulation of muscle contraction
mitochondrion organization
mesoderm development
myofibril assembly
skeletal muscle thin filament assembly
sarcomere organization
muscle cell cellular homeostasis
muscle organ morphogenesis
calcium ion binding
tropomyosin binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0