This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01827973_m1
See other AGPAT1 GE Assays ›
Gene Symbol:
Agpat1
Gene Name:
1-Acylglycerol-3-phosphate O-acyltransferase 1
Gene Aliases:
AGPAT, AGPAT1, AGPAT2, CG3812, Dmel\CG3812, Dmel_CG3812, dAGPAT1
Chromosome Location:
Chr.X: 12765266 - 12772851 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0030421
Assay ID Dm01827973_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Agpat1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001272565.1 NP_001259494.1 2 - 3 496 75
NM_132600.2 NP_572828.1 2 - 3 634 75
NM_167337.2 NP_727641.1 2 - 3 725 75
GenBank mRNA AY118588.1 - 2 - 3 573 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

phosphatidic acid biosynthetic process
1-acylglycerol-3-phosphate O-acyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0