This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01826523_g1
See other CAMP GE Assays ›
Gene Symbol:
Camp
Gene Name:
Cathelicidin-like antimicrobial protein
Gene Aliases:
CG6568, Dmel\CG6568, Dmel_CG6568, cg6568, dCAMP
Chromosome Location:
Chr.2R: 17405304 - 17407427 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0034210
Assay ID Dm01826523_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Camp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001299594.1 NP_001286523.1 1 - 2 281 74
NM_137360.2 NP_611204.2 2 - 3 373 74
GenBank mRNA AY121689.1 - 2 - 3 371 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Drosophila melanogaster

Gene Ontology Categories:

Function(s)

defense response to bacterium

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0