This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01826374_g1
See other UNC-104 GE Assays ›
Gene Symbol:
unc-104
Gene Name:
uncoordinated-104
Gene Aliases:
CG8566, DUNC104, DUnc-104, DmCG8566, DmDUnc104, DmKlp53D, Dmel\CG8566, Dmel_CG8566, Dunc104, Imac, KIF 1B, KIF1A, KIF1A/1B, Kif1a, Klp53D, Unc-104, Unc104, bris, imac, unc104
Chromosome Location:
Chr.2R: 16751503 - 16772671 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0267002
Assay ID Dm01826374_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: unc-104
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001103876.3 NP_001097346.1 25 - 26 5304 63
NM_001259444.2 NP_001246373.1 24 - 25 5277 63
NM_001259445.2 NP_001246374.1 25 - 26 5295 63
NM_001274104.2 NP_001261033.1 22 - 23 5077 63
NM_001316424.1 NP_001303353.1 22 - 23 5077 63
NM_137311.5 NP_611155.3 25 - 26 5469 63
NM_166190.4 NP_725607.2 24 - 25 5262 63
NM_166192.4 NP_725610.2 23 - 24 5271 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

microtubule binding motor protein

Gene Ontology Categories:

Function(s) Process(es)

microtubule-based movement
axon guidance
neuromuscular junction development
axo-dendritic transport
anterograde axonal transport
larval locomotory behavior
synaptic vesicle maturation
vesicle-mediated transport
cytoskeleton-dependent intracellular transport
positive regulation of insulin secretion
regulation of locomotion
negative regulation of synaptic growth at neuromuscular junction
filopodium assembly
vesicle transport along microtubule
synaptic vesicle transport
anterograde synaptic vesicle transport
regulation of dendrite morphogenesis
regulation of synapse assembly
microtubule motor activity
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
ATPase activity
GTP-dependent protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0