This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01826189_g1
See other NOX GE Assays ›
Gene Symbol:
Nox
Gene Name:
NADPH oxidase
Gene Aliases:
CG15924, CG34399, CG3896, DmNox, Dmel\CG34399, Dmel_CG34399, NOX, NOX-1, Nox1, dNox, dnox
Chromosome Location:
Chr.2R: 16307269 - 16316485 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0085428
Assay ID Dm01826189_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nox
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001103866.2 NP_001097336.1 9 - 10 3655 100
NM_001299557.1 NP_001286486.1 9 - 10 5535 100
NM_137272.2 NP_611116.2 9 - 10 3862 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Drosophila melanogaster

Panther Classification:

Molecular Function -

oxidase

Gene Ontology Categories:

Function(s) Process(es)

ovulation
iron assimilation by reduction and transport
positive regulation of smooth muscle contraction
positive regulation of calcium-mediated signaling
oxidation-reduction process
ferric-chelate reductase activity
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0