This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01825653_g1
See other SYD GE Assays ›
Gene Symbol:
syd
Gene Name:
sunday driver
Gene Aliases:
CG8110, Dmel\CG8110, Dmel_CG8110, JIP-3, Syd, dsyd-1, jip3
Chromosome Location:
Chr.3L: 7916650 - 7936120 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0024187
Assay ID Dm01825653_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: syd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001169902.2 NP_001163373.1 4 - 5 1149 66
NM_001169903.1 NP_001163374.1 4 - 5 574 66
NM_001274619.1 NP_001261548.1 3 - 4 1128 66
NM_001274620.1 NP_001261549.1 3 - 4 1128 66
NM_001274621.1 NP_001261550.1 3 - 4 1128 66
NM_001274622.1 NP_001261551.1 3 - 4 1128 66
NM_001274623.1 NP_001261552.1 3 - 4 1128 66
NM_001274624.1 NP_001261553.1 3 - 4 1128 66
NM_001300046.1 NP_001286975.1 3 - 4 1128 66
NM_079913.3 NP_524652.1 4 - 5 1149 66
NM_168245.3 NP_729311.2 4 - 5 574 66
GenBank mRNA AF262045.1 - 4 - 5 903 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

activation of JUN kinase activity
axo-dendritic transport
vesicle-mediated transport
regulation of JNK cascade
MAP-kinase scaffold activity
JUN kinase binding
kinesin binding
receptor signaling complex scaffold activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0