This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01824601_g1
See other CBP80 GE Assays ›
Gene Symbol:
Cbp80
Gene Name:
cap binding protein 80
Gene Aliases:
CBP80, CG7035, Dmel\CG7035, Dmel_CG7035, EG:84H4.3, cbp80
Chromosome Location:
Chr.X: 4650015 - 4658259 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0022942
Assay ID Dm01824601_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cbp80
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080011.3 NP_524750.2 5 - 6 2380 67
NM_167012.3 NP_726938.1 5 - 6 2415 67
GenBank mRNA AY051796.1 - 5 - 6 2355 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Panther Classification:

Molecular Function -

RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
mRNA splicing, via spliceosome
RNA interference
production of siRNA involved in RNA interference
primary miRNA processing
gene silencing by miRNA
negative regulation of viral genome replication
mRNA cis splicing, via spliceosome
mRNA transport
RNA cap binding
mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0