This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01822965_g1
See other TSG101 GE Assays ›
Gene Symbol:
TSG101
Gene Name:
Tumor susceptibility gene 101
Gene Aliases:
CG9712, DmTSG101, Dmel\CG9712, Dmel_CG9712, ESCRT-I, Ept, Tsg1-1, Tsg101, Vps23, burs, dTSg101, dTsg101, dVps23, ept, tsg101, vps23
Chromosome Location:
Chr.3L: 16842294 - 16844585 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0036666
Assay ID Dm01822965_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TSG101
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_079396.4 NP_524120.1 2 - 3 467 74
GenBank mRNA AF315343.1 - 2 - 3 174 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
terminal branching, open tracheal system
protein transport
neuron remodeling
liquid clearance, open tracheal system
maintenance of apical/basal cell polarity
multivesicular body assembly
negative regulation of imaginal disc growth
protein binding
ubiquitin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header