This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01822868_g1
See other PKA-C3 GE Assays ›
Gene Symbol:
Pka-C3
Gene Name:
Protein kinase, cAMP-dependent, catalytic subunit 3
Gene Aliases:
5-23, CG6117, DC2, Dmel\CG6117, Dmel_CG6117, Dpck, PKA, PKA C3, PKA-C-3, PKA-C3, PKAC3, Pk72A, Pk?5, pka-C3
Chromosome Location:
Chr.3L: 15925805 - 15954797 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0000489
Assay ID Dm01822868_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pka-C3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_079373.4 NP_524097.2 6 - 7 2272 103
NM_168638.3 NP_730083.2 7 - 8 1590 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Drosophila melanogaster

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
signal transduction
imaginal disc-derived wing morphogenesis
peptidyl-serine phosphorylation
cAMP-dependent protein kinase activity
protein binding
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0