This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01820758_g1
See other 4E-T GE Assays ›
Gene Symbol:
4E-T
Gene Name:
eIF4E-Transporter
Gene Aliases:
CG11097, CG11232, CG32016, Dm4E-T, Dmel\CG32016, Dmel_CG32016
Chromosome Location:
Chr.4: 951542 - 958392 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0052016
Assay ID Dm01820758_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4E-T
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001272149.2 NP_001259078.1 7 - 8 2842 63
NM_166796.2 NP_726618.3 8 - 9 2992 63
NM_166797.2 NP_726619.2 8 - 9 2967 63
NM_166798.2 NP_726620.2 7 - 8 3157 63
GenBank mRNA BT001473.1 - 7 - 8 3157 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Drosophila melanogaster

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of translation
negative regulation of eukaryotic translation initiation factor 4F complex assembly
mRNA binding
eukaryotic initiation factor 4E binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0