This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01820560_g1
See other CRK GE Assays ›
Gene Symbol:
Crk
Gene Name:
Crk oncogene
Gene Aliases:
CG1587, CRK, D-CRK, D-Crk, DCrk, Dcrk, Dmel\CG1587, Dmel_CG1587, crk, dCRK, dCrk
Chromosome Location:
Chr.4: 210224 - 213165 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0024811
Assay ID Dm01820560_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crk
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001258482.2 NP_001245411.1 5 - 6 797 86
NM_001258483.2 NP_001245412.1 6 - 7 981 86
NM_001297781.1 NP_001284710.1 3 - 4 487 86
NM_001297782.1 NP_001284711.1 5 - 6 541 86
NM_143651.6 NP_651908.1 5 - 6 821 86
NM_166743.3 NP_726549.1 6 - 7 746 86
NM_166744.3 NP_726550.1 5 - 6 767 86
GenBank mRNA AF112976.1 - 5 - 6 731 86
AY061094.1 - 5 - 6 649 86
BT012456.1 - 5 - 6 819 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

phagocytosis, engulfment
border follicle cell migration
myoblast fusion
regulation of actin cytoskeleton organization
regulation of GTPase activity
positive regulation of JNK cascade
imaginal disc fusion, thorax closure
vascular endothelial growth factor receptor signaling pathway
ephrin receptor signaling pathway
receptor tyrosine kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0