This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01819276_g1
See other CAF1-105 GE Assays ›
Gene Symbol:
Caf1-105
Gene Name:
Chromatin assembly factor 1, p105 subunit
Gene Aliases:
CAF p105, CAF1, CAF1-105, CAF1-p105, CAF1-p75, CAF1-p75/p105, CAF105/75, CAF1p105, CAF1p75, CG12892, Caf-105, Caf1, Caf1-10, Dmel\CG12892, Dmel_CG12892, P105, caf1-105, dCAF-1-p105, dCAF1-p105, dCAF1-p75, p105, p105/p75, p75, p75/p105
Chromosome Location:
Chr.2R: 10466001 - 10468540 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0033526
Assay ID Dm01819276_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Caf1-105
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_136745.5 NP_610589.2 1 - 2 683 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
histone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header